Anti-PSMD3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA312061EA 656 CHF
ORIGTA312061
Anti-PSMD3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: PSMD3 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR