Anti-PRKCZ Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312044EA 656 CHF
ORIGTA312044
Anti-PRKCZ Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-3000; Immunochemistry: 1:50-100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: PRKCZ (protein kinase C, zeta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR