Anti-MAD2L1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA311990EA 656 CHF
ORIGTA311990
Anti-MAD2L1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: MAD2L1 (MAD2 mitotic arrest deficient-like 1 (yeast))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR