Anti-HNRNPK Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA311956EA 656 CHF
ORIGTA311956
Anti-HNRNPK Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: HNRNPK (heterogeneous nuclear ribonucleoprotein K)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR