Anti-C1S Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA311912EA 632 CHF
ORIGTA311912
Anti-C1S Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: C1S (complement component 1, s subcomponent)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR