Anti-IFIT1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA311892EA 642 CHF
ORIGTA311892
Anti-IFIT1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: IFIT1 (interferon-induced protein with tetratricopeptide repeats 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR