Anti-CYP2C8 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA311802EA 656 CHF
ORIGTA311802
Anti-CYP2C8 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: CYP2C8 (cytochrome P450, family 2, subfamily C, polypeptide 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR