Anti-ADORA2A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA311793EA 642 CHF
ORIGTA311793
Anti-ADORA2A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: ADORA2A (adenosine A2a receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR