Anti-GNRHR Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA311775EA 642 CHF
ORIGTA311775
Anti-GNRHR Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: GNRHR (gonadotropin-releasing hormone receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR