Anti-FGFR1OP Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA311655EA 619 CHF
ORIGTA311655
Anti-FGFR1OP Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: FGFR1OP (FGFR1 oncogene partner)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR