Anti-LRRK2 Goat Antibody

Supplier: OriGene
ORIGTA311617EA 624 CHF
ORIGTA311617
Anti-LRRK2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.3-1?g/ml; Immunochemistry: 1-2?g/ml; Immunofluorescence: customer reported positive

Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR