Anti-KLHDC8B Goat Antibody

Supplier: OriGene
ORIGTA311594EA 627 CHF
ORIGTA311594
Anti-KLHDC8B Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: KLHDC8B (kelch domain containing 8B)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR