Anti-TRPV3 Goat Antibody

Supplier: OriGene
ORIGTA311578EA 624 CHF
ORIGTA311578
Anti-TRPV3 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-1?g/ml

Type: Primary
Antigen: TRPV3 (transient receptor potential cation channel, subfamily V, member 3)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR