Anti-PEBP4 Goat Antibody

Supplier: OriGene
ORIGTA311576EA 624 CHF
ORIGTA311576
Anti-PEBP4 Goat Antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Western Blot: 1-3?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: PEBP4 (phosphatidylethanolamine-binding protein 4)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR