Anti-DYDC1 Goat Antibody

Supplier: OriGene
ORIGTA311571EA 640 CHF
ORIGTA311571
Anti-DYDC1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.1-1?g/ml

Type: Primary
Antigen: DYDC1 (DPY30 domain containing 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR