Anti-UBE2F Goat Antibody

Lieferant: OriGene
ORIGTA311564EA 637 CHF
ORIGTA311564
Anti-UBE2F Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.05-0.2?g/ml

Type: Primary
Antigen: UBE2F (ubiquitin-conjugating enzyme E2F (putative))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR