Anti-FUBP1 Goat Antibody

Lieferant: OriGene
ORIGTA311561EA 637 CHF
ORIGTA311561
Anti-FUBP1 Goat Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: 0.03-0.1?g/ml; Immunochemistry: 4-8?g/ml

Type: Primary
Antigen: FUBP1 (far upstream element (FUSE) binding protein 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR