Anti-SNX18 Goat Antibody

Supplier: OriGene
ORIGTA311557EA 624 CHF
ORIGTA311557
Anti-SNX18 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: SNX18 (sorting nexin 18)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR