Anti-CREB3L1 Goat Antibody

Lieferant: OriGene
ORIGTA311556EA 640 CHF
ORIGTA311556
Anti-CREB3L1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: CREB3L1 (cAMP responsive element binding protein 3-like 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR