Anti-TMEM205 Goat Antibody

Supplier: OriGene
ORIGTA311555EA 627 CHF
ORIGTA311555
Anti-TMEM205 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.03-0.1?g/ml

Type: Primary
Antigen: TMEM205 (transmembrane protein 205)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR