Anti-SLC4A11 Goat Antibody

Lieferant: OriGene
ORIGTA311544EA 637 CHF
ORIGTA311544
Anti-SLC4A11 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: SLC4A11 (solute carrier family 4, sodium borate transporter, member 11)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR