Anti-NOX5 Goat Antibody

Supplier: OriGene
ORIGTA311523EA 637 CHF
ORIGTA311523
Anti-NOX5 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: NOX5 (NADPH oxidase, EF-hand calcium binding domain 5)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR