Anti-PLA2G4A Goat Antibody

Supplier: OriGene
ORIGTA311521EA 637 CHF
ORIGTA311521
Anti-PLA2G4A Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: PLA2G4A (phospholipase A2, group IVA (cytosolic, calcium-dependent))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR