Anti-SLC26A6 Goat Antibody

Lieferant: OriGene
ORIGTA311513EA 640 CHF
ORIGTA311513
Anti-SLC26A6 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 10?g/ml; Immunochemistry: 0.1-0.3mg/ml

Type: Primary
Antigen: SLC26A6 (solute carrier family 26 (anion exchanger), member 6)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR