Anti-RPS6KA2 Goat Antibody

Supplier: OriGene
ORIGTA311500EA 637 CHF
ORIGTA311500
Anti-RPS6KA2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.01-0.03?g/ml

Type: Primary
Antigen: RPS6KA2 (ribosomal protein S6 kinase, 90kDa, polypeptide 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR