Anti-PMAIP1 Goat Antibody

Lieferant: OriGene
ORIGTA311499EA 637 CHF
ORIGTA311499
Anti-PMAIP1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.03-0.1?g/ml

Type: Primary
Antigen: PMAIP1 (phorbol-12-myristate-13-acetate-induced protein 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR