Anti-HCN3 Goat Antibody

Lieferant: OriGene
ORIGTA311496EA 637 CHF
ORIGTA311496
Anti-HCN3 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.01-0.03?g/ml

Type: Primary
Antigen: HCN3 (hyperpolarization activated cyclic nucleotide-gated potassium channel 3)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR