Anti-DCUN1D1 Goat Antibody

Supplier: OriGene
ORIGTA311491EA 624 CHF
ORIGTA311491
Anti-DCUN1D1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.3-1?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: DCUN1D1 (DCN1, defective in cullin neddylation 1, domain containing 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR