Anti-NUP107 Goat Antibody

Lieferant: OriGene
ORIGTA311490EA 637 CHF
ORIGTA311490
Anti-NUP107 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: NUP107 (nucleoporin 107kDa)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR