Anti-PRMT7 Goat Antibody

Supplier: OriGene
ORIGTA311482EA 624 CHF
ORIGTA311482
Anti-PRMT7 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: PRMT7 (protein arginine methyltransferase 7)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR