Anti-RIF1 Goat Antibody

Supplier: OriGene
ORIGTA311474EA 624 CHF
ORIGTA311474
Anti-RIF1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: RIF1 (RAP1 interacting factor homolog (yeast))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR