Anti-NODAL Goat Antibody

Supplier: OriGene
ORIGTA311471EA 640 CHF
ORIGTA311471
Anti-NODAL Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-2?g/ml

Type: Primary
Antigen: NODAL (nodal growth differentiation factor)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR