Anti-FBXL12 Goat Antibody

Lieferant: OriGene
ORIGTA311466EA 640 CHF
ORIGTA311466
Anti-FBXL12 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: FBXL12 (F-box and leucine-rich repeat protein 12)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR