Anti-RHCG Goat Antibody

Supplier: OriGene
ORIGTA311456EA 624 CHF
ORIGTA311456
Anti-RHCG Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: RHCG (Rh family, C glycoprotein)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR