Anti-SGSM3 Goat Antibody

Supplier: OriGene
ORIGTA311446EA 624 CHF
ORIGTA311446
Anti-SGSM3 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 2-3?g/ml

Type: Primary
Antigen: SGSM3 (small G protein signaling modulator 3)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR