Anti-IRF2BP1 Goat Antibody

Supplier: OriGene
ORIGTA311443EA 627 CHF
ORIGTA311443
Anti-IRF2BP1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml

Type: Primary
Antigen: IRF2BP1 (interferon regulatory factor 2 binding protein 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR