Anti-MLL4 Goat Antibody

Supplier: OriGene
ORIGTA311425EA 637 CHF
ORIGTA311425
Anti-MLL4 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: MLL4
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR