Anti-SIRT1 Goat Antibody

Supplier: OriGene
ORIGTA311399EA 624 CHF
ORIGTA311399
Anti-SIRT1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.05-0.2?g/ml; Immunochemistry: 3-5?g/ml

Type: Primary
Antigen: SIRT1 (sirtuin 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR