Anti-IRS4 Goat Antibody

Supplier: OriGene
ORIGTA311395EA 637 CHF
ORIGTA311395
Anti-IRS4 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.2-0.6?g/ml

Type: Primary
Antigen: IRS4 (insulin receptor substrate 4)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR