Anti-PROL1 Goat Antibody

Supplier: OriGene
ORIGTA311390EA 627 CHF
ORIGTA311390
Anti-PROL1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: PROL1 (proline rich, lacrimal 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR