Anti-LGALS1 Goat Antibody

Supplier: OriGene
ORIGTA311389EA 637 CHF
ORIGTA311389
Anti-LGALS1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: LGALS1 (lectin, galactoside-binding, soluble, 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR