Anti-PLA2R1 Goat Antibody

Supplier: OriGene
ORIGTA311382EA 637 CHF
ORIGTA311382
Anti-PLA2R1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: PLA2R1 (phospholipase A2 receptor 1, 180kDa)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR