Anti-IGF2BP1 Goat Antibody

Supplier: OriGene
ORIGTA311367EA 624 CHF
ORIGTA311367
Anti-IGF2BP1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: IGF2BP1 (insulin-like growth factor 2 mRNA binding protein 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR