Anti-POU3F3 Goat Antibody

Supplier: OriGene
ORIGTA311357EA 624 CHF
ORIGTA311357
Anti-POU3F3 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: POU3F3 (POU class 3 homeobox 3)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR