Anti-YAP1 Goat Antibody

Supplier: OriGene
ORIGTA311348EA 624 CHF
ORIGTA311348
Anti-YAP1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: YAP1 (Yes-associated protein 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR