Anti-NOD1 Goat Antibody

Supplier: OriGene
ORIGTA311347EA 624 CHF
ORIGTA311347
Anti-NOD1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-1.5?g/ml

Type: Primary
Antigen: NOD1 (nucleotide-binding oligomerization domain containing 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR