Anti-NET1 Goat Antibody

Supplier: OriGene
ORIGTA311345EA 624 CHF
ORIGTA311345
Anti-NET1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.3?g/ml; Immunochemistry: 2.5?g/ml

Type: Primary
Antigen: NET1 (neuroepithelial cell transforming 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR