Anti-LCK Goat Antibody

Supplier: OriGene
ORIGTA311330EA 637 CHF
ORIGTA311330
Anti-LCK Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml

Type: Primary
Antigen: LCK (lymphocyte-specific protein tyrosine kinase)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR