Anti-GPM6A Goat Antibody

Lieferant: OriGene
ORIGTA311328EA 637 CHF
ORIGTA311328
Anti-GPM6A Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: GPM6A (glycoprotein M6A)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR