Anti-GEM Goat Antibody

Supplier: OriGene
ORIGTA311327EA 627 CHF
ORIGTA311327
Anti-GEM Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: GEM (GTP binding protein overexpressed in skeletal muscle)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR